ENA447958500 Enamine

methyl 1-(2-aminopropanoyl)piperidine-3-carboxylate hydrochloride

  • MDL number MFCD28397651
  • Cas Number : 1593136-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209298
Availability details for
Quantity
  • Availability
  • Add to Cart