ENA447958504 Enamine

5-(methoxymethyl)-1,2-oxazole-3-carbaldehyde

  • MDL number MFCD06804221
  • Cas Number : 893749-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209308
Availability details for
Quantity
  • Availability
  • Add to Cart