ENA447958512 Enamine

2-fluoro-6-hydrazinylpyridine

  • MDL number MFCD00214745
  • Cas Number : 80714-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209317
Availability details for
Quantity
  • Availability
  • Add to Cart