ENA447958519 Enamine

{[1,1'-biphenyl]-4-yl}(oxolan-2-yl)methanol

  • MDL number MFCD16357164
  • Cas Number : 1285673-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208260
Availability details for
Quantity
  • Availability
  • Add to Cart