ENA447958535 Enamine

4-methyl-2-phenylpyrrolidine

  • MDL number MFCD05215075
  • Cas Number : 891763-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208316
Availability details for
Quantity
  • Availability
  • Add to Cart