ENA447958538 Enamine

2-phenylimidazo[1,2-a]pyridin-6-amine

  • MDL number MFCD05863414
  • Cas Number : 887576-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209375
Availability details for
Quantity
  • Availability
  • Add to Cart