ENA447958544 Enamine

3-[(2,4-dichlorophenoxy)methyl]-1H-1,2,4-triazole-5-carboxamide

  • MDL number MFCD01936204
  • Cas Number : 400080-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209384
Availability details for
Quantity
  • Availability
  • Add to Cart