ENA447958548 Enamine

4-(azepan-1-yl)-2-methylaniline

  • MDL number MFCD11848936
  • Cas Number : 847455-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208623
Availability details for
Quantity
  • Availability
  • Add to Cart