ENA447958550 Enamine

3-(3,4-difluorophenyl)prop-2-enal

  • Cas Number : 1334528-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208626
Availability details for
Quantity
  • Availability
  • Add to Cart