ENA447958558 Enamine

1-benzyl-1H-1,2,3-triazole-5-carboxylic acid

  • MDL number MFCD11557215
  • Cas Number : 103264-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208643
Availability details for
Quantity
  • Availability
  • Add to Cart