ENA447958563 Enamine

(2R)-2-methyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD28397584
  • Cas Number : 351458-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-44
Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208652
Availability details for
Quantity
  • Availability
  • Add to Cart