ENA447958565 Enamine

2-(aminomethyl)butan-1-amine

  • MDL number MFCD21183657
  • Cas Number : 57312-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209479
Availability details for
Quantity
  • Availability
  • Add to Cart