ENA447958566 Enamine

methyl 3-cyclohexyl-2-hydroxypropanoate

  • MDL number MFCD21331230
  • Cas Number : 1033777-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209480
Availability details for
Quantity
  • Availability
  • Add to Cart