ENA447958567 Enamine

(2-aminoethyl)[2-(piperidin-1-yl)ethyl]amine

  • MDL number MFCD00053444
  • Cas Number : 5625-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209481
Availability details for
Quantity
  • Availability
  • Add to Cart