ENA447958569 Enamine

[3-({2-[(2-chloroethyl)amino]ethyl}amino)propoxy]phosphonic acid dihydrochloride

  • MDL number MFCD01757095
  • Cas Number : 158401-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208655
Availability details for
Quantity
  • Availability
  • Add to Cart