ENA447958572 Enamine

2-{3-amino-8-azabicyclo[3.2.1]octan-8-yl}-N,N-dimethylacetamide

  • MDL number MFCD14602686
  • Cas Number : 1248379-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209487
Availability details for
Quantity
  • Availability
  • Add to Cart