ENA447958573 Enamine

3-chloro-2-(trimethylsilyl)pyridine

  • MDL number MFCD08064046
  • Cas Number : 79698-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209492
Availability details for
Quantity
  • Availability
  • Add to Cart