ENA447958574 Enamine

3-(4-aminophenoxy)propan-1-ol

  • MDL number MFCD11205805
  • Cas Number : 99190-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209494
Availability details for
Quantity
  • Availability
  • Add to Cart