ENA447958587 Enamine

2-(methanesulfonylmethyl)phenol

  • MDL number MFCD24721304
  • Cas Number : 53380-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209527
Availability details for
Quantity
  • Availability
  • Add to Cart