ENA447958588 Enamine

N-(2-aminoethyl)-4-chloro-2-methylbenzene-1-sulfonamide hydrochloride

  • MDL number MFCD28397671
  • Cas Number : 1603725-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209528
Availability details for
Quantity
  • Availability
  • Add to Cart