ENA447958593 Enamine

5-methoxypyrimidin-4-amine

  • MDL number MFCD01646126
  • Cas Number : 695-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-119
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209533
Availability details for
Quantity
  • Availability
  • Add to Cart