ENA447958595 Enamine

methyl 3-amino-4-(4-methylphenyl)thiophene-2-carboxylate

  • MDL number MFCD04116501
  • Cas Number : 160133-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209535
Availability details for
Quantity
  • Availability
  • Add to Cart