ENA447958599 Enamine

(1-cyanoethyl)urea

  • MDL number MFCD12793054
  • Cas Number : 1250694-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208690
Availability details for
Quantity
  • Availability
  • Add to Cart