ENA447958602 Enamine

2-(2-hydroxyphenyl)-4,5-dihydro-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD01191410
  • Cas Number : 49608-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209566
Availability details for
Quantity
  • Availability
  • Add to Cart