ENA447958603 Enamine

1-(4-cyano-2-fluorophenyl)piperidine-4-carboxylic acid

  • MDL number MFCD11038721
  • Cas Number : 1156886-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209569
Availability details for
Quantity
  • Availability
  • Add to Cart