ENA447958606 Enamine

3-iodo-5-methylaniline

  • MDL number MFCD12965021
  • Cas Number : 74586-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209574
Availability details for
Quantity
  • Availability
  • Add to Cart