ENA447958607 Enamine

3,3,3-trifluoro-N-[(piperidin-3-yl)methyl]propanamide hydrochloride

  • MDL number MFCD28397554
  • Cas Number : 1582121-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208347
Availability details for
Quantity
  • Availability
  • Add to Cart