ENA447958611 Enamine

3-hydroxy-4-methyl-2,5-dihydrofuran-2-one

  • MDL number MFCD15145816
  • Cas Number : 22438-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209599
Availability details for
Quantity
  • Availability
  • Add to Cart