ENA447958612 Enamine

2-fluoro-5-methylbenzene-1-sulfonamide

  • MDL number MFCD00462129
  • Cas Number : 438539-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209600
Availability details for
Quantity
  • Availability
  • Add to Cart