ENA447958613 Enamine

methyl 2-(2-amino-4,5-dihydro-1,3-thiazol-4-yl)acetate

  • MDL number MFCD02656566
  • Cas Number : 103541-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209602
Availability details for
Quantity
  • Availability
  • Add to Cart