ENA447958614 Enamine

2-amino-4-methoxy-2-methylbutanenitrile

  • MDL number MFCD16127694
  • Cas Number : 1249634-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209605
Availability details for
Quantity
  • Availability
  • Add to Cart