ENA447958616 Enamine

methyl 6-fluoro-2-oxo-2,3-dihydro-1H-indole-5-carboxylate

  • MDL number MFCD24617111
  • Cas Number : 1265965-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209607
Availability details for
Quantity
  • Availability
  • Add to Cart