ENA447958618 Enamine

4-(trifluoromethoxy)pyridin-2-ol

  • MDL number MFCD25450566
  • Cas Number : 1361737-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208248
Availability details for
Quantity
  • Availability
  • Add to Cart