ENA447958619 Enamine

4-[(1H-imidazol-2-yl)methyl]piperidine dihydrochloride

  • MDL number MFCD09909009
  • Cas Number : 90747-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209619
Availability details for
Quantity
  • Availability
  • Add to Cart