ENA447958622 Enamine

2-bromobutyl acetate

  • MDL number MFCD20626738
  • Cas Number : 13333-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208750
Availability details for
Quantity
  • Availability
  • Add to Cart