ENA447958624 Enamine

2-(3-chlorophenyl)ethane-1-sulfonamide

  • MDL number MFCD26064735
  • Cas Number : 919353-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209625
Availability details for
Quantity
  • Availability
  • Add to Cart