ENA447958625 Enamine

2-methyl-1-oxo-1,2-dihydroisoquinoline-3-carboxylic acid

  • MDL number MFCD06368805
  • Cas Number : 89806-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238-240
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209626
Availability details for
Quantity
  • Availability
  • Add to Cart