ENA447958626 Enamine

5,6-dichloro-3-imino-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD25962616
  • Cas Number : 35190-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209627
Availability details for
Quantity
  • Availability
  • Add to Cart