ENA447958627 Enamine

1-bromo-2-chloro-3-fluoro-4-(trifluoromethyl)benzene

  • MDL number MFCD13195463
  • Cas Number : 1260890-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208752

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208752
Availability details for
Quantity
  • Availability
  • Add to Cart