ENA447958628 Enamine

1-(4-chloro-1H-pyrrole-2-carbonyl)piperidin-4-amine hydrochloride

  • MDL number MFCD28397682
  • Cas Number : 1606501-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209629
Availability details for
Quantity
  • Availability
  • Add to Cart