ENA447958630 Enamine

ethyl 5-(chlorosulfonyl)-2-methylbenzoate

  • MDL number MFCD11057847
  • Cas Number : 924859-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208755
Availability details for
Quantity
  • Availability
  • Add to Cart