ENA447958634 Enamine

methyl({[4-(2,2,2-trifluoroethoxy)phenyl]methyl})amine

  • MDL number MFCD11629897
  • Cas Number : 1095157-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208757
Availability details for
Quantity
  • Availability
  • Add to Cart