ENA447958635 Enamine

1-[(4-chlorophenyl)methyl]-3-(trifluoromethyl)-1H-pyrazol-5-ol

  • MDL number MFCD23175287
  • Cas Number : 1803606-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209640
Availability details for
Quantity
  • Availability
  • Add to Cart