ENA447958636 Enamine

3,4-dimethyl thiophene-3,4-dicarboxylate

  • MDL number MFCD01233039
  • Cas Number : 4282-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209642
Availability details for
Quantity
  • Availability
  • Add to Cart