ENA447958638 Enamine

1-(azepan-1-yl)-2-(ethylamino)ethan-1-one

  • MDL number MFCD11129202
  • Cas Number : 97454-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209644
Availability details for
Quantity
  • Availability
  • Add to Cart