ENA447958639 Enamine

methyl 3-amino-2-(methylamino)pyridine-4-carboxylate

  • MDL number MFCD21288016
  • Cas Number : 1465010-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209645
Availability details for
Quantity
  • Availability
  • Add to Cart