ENA447958640 Enamine

2-benzoyl-4-fluorophenol

  • MDL number MFCD03094354
  • Cas Number : 362-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208758
Availability details for
Quantity
  • Availability
  • Add to Cart