ENA447958644 Enamine

ethynylcycloheptane

  • MDL number MFCD00041702
  • Cas Number : 783339-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209652
Availability details for
Quantity
  • Availability
  • Add to Cart