ENA447958646 Enamine

2-(2-aminoethoxy)ethane-1-sulfonamide hydrochloride

  • MDL number MFCD28397688
  • Cas Number : 1803565-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209657
Availability details for
Quantity
  • Availability
  • Add to Cart