ENA447958647 Enamine

1-(5-amino-2-fluorophenyl)-3-methylurea

  • MDL number MFCD24262148
  • Cas Number : 1546093-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208761
Availability details for
Quantity
  • Availability
  • Add to Cart